Learn More
Medchemexpress LLC (S)-Ofloxacin-d3 | 2208780-63-4 | 98.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0330S11MG
Encompass_Preferred
(S)-Ofloxacin-d3 is a deuterium-labeled version of Levofloxacin, a synthetic fluoroquinolone. It acts as an antibacterial agent by inhibiting bacterial DNA gyrase, which stops DNA replication.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS
  • Contains stable heavy isotopes for quantitation during drug development
  • Potential to influence pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-06764