Learn More
Medchemexpress LLC (S)-Omeprazole (Standard) | 119141-88-7 | 345.42 | 50 MG
SDP
Catalog No. 5000386051
Encompass_Preferred
Esomeprazole (Standard) is intended for use as a laboratory chemical in the manufacture of substances. This product is for research use only and should be handled by suitably qualified experienced scientists in appropriately equipped and authorized facilities. It has not been fully validated for medical applications.
  • Stable under recommended storage conditions.
  • Should be stored at -20°C, sealed, away from light and moisture.
  • Ships at room temperature if less than 2 weeks.
  • Appears as a solid.
Catalog No. 50-003-86051 Supplier Medchemexpress LLC Supplier No. HY17021R50MG
Esomeprazole (Standard) is intended for use as a laboratory chemical in the manufacture of substances. This product is for research use only and should be handled by suitably qualified experienced scientists in appropriately equipped and authorized facilities. It has not been fully validated for medical applications.
  • Stable under recommended storage conditions.
  • Should be stored at -20°C, sealed, away from light and moisture.
  • Ships at room temperature if less than 2 weeks.
  • Appears as a solid.