Learn More
Zymo Research Corporation S&P 5X Digestion Buffer, 32 ml
SDP
Catalog No. 502037065
Encompass_Preferred
The S&P 5X Digestion Buffer is designed to be used in the Quick-cfDNA Serum & Plasma Kit. It is a concentrated Proteinase K digestion buffer and facilities efficient DNA recovery from serum, plasma, and other protein-rich samples.
Catalog No. 50-203-7065 Supplier Zymo Research Corporation Supplier No. D4076132
The S&P 5X Digestion Buffer is designed to be used in the Quick-cfDNA Serum & Plasma Kit. It is a concentrated Proteinase K digestion buffer and facilities efficient DNA recovery from serum, plasma, and other protein-rich samples.