Learn More
Cayman Chemical S-p38 MAPK InhIbItr III 10mg
SDP
Catalog No. 502982894
Encompass_Preferred
A methylsulfanylimidazole that inhibits p38 MAP kinase (IC50 = 0.90 µM in vitro); potently blocks the release of TNF-α and IL-1β from human peripheral blood mononuclear cells (IC50s = 0.37 and 0.044 µM, respectively)
Catalog No. 50-298-2894 Supplier Cayman Chemical Supplier No. 1969610 MG
A methylsulfanylimidazole that inhibits p38 MAP kinase (IC50 = 0.90 µM in vitro); potently blocks the release of TNF-α and IL-1β from human peripheral blood mononuclear cells (IC50s = 0.37 and 0.044 µM, respectively)