Learn More
AdipoGen (S)-Perharidine 1 (5 mg)
SDP
Supplier:  AdipoGen AGMRC0012M005
Encompass_Preferred
Chemical. CAS: 1133437-81-6. Formula: C25H30N6O. MW: 430.6. Potent and selective inhibitor of cyclin dependent kinases CDK1, CDK2, CDK5 and CDK9. Apoptosis inducer. Inhibits the proliferation of various cancer cell lines. Casein kinase 1 (CK1delta/epsilon) and DYRK1A inhibitor. Potential anti-inflammatory compound. Potential antidiabetic compound.
Catalog No. 50-205-4227