Learn More
Medchemexpress LLC HY-116514 100mg Medchemexpress, (S)-(−)-Perillyl alcohol CAS:18457-55-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY116514100MG
Encompass_Preferred
Medchemexpress, HY-116514 100mg (S)-(−)-Perillyl alcohol CAS:18457-55-1 (S)-(−)-Perillyl alcohol is a monoterpene found in lavender, inhibits farnesylation of Ras, upregulates the mannose-6-phosphate receptor and induces apoptosis . Anti-cancer activity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2511