Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(-)-Perillyl Alcohol

Catalog No. 21839150G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-839-150G 50 g
21-839-110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 21-839-150G Supplier Sigma Aldrich Supplier No. 21839150G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(-)-Perillyl alcohol is a monoterpenoid compound found in the essential oils of cherries, lavender and spearmint. It shows potent anticancer activity.(S)-(-)-Perillyl alcohol (POH or 4-isopropenyl cyclohexene carbinol) can be used as a starting material in the synthesis of: Perillyl alcohol neoglycoside derivatives as potential anti cancer agents Amino-modified derivatives of (S)-perillyl alcohol as potent antIpr∗

Specifications

CAS 18457-55-1
Density 0.96 g/mL at 25°C
Boiling Point 119°C to 121°C (11 mmHg, lit.)
Molecular Formula C10H16O
Refractive Index n20/D 1.501
Linear Formula C10H16O
MDL Number MFCD00062995
Quantity 50 g
Synonym p-Mentha-1,8-diene-7-ol
Molecular Weight (g/mol) 152.23 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.