Learn More
TARGETMOL CHEMICALS INC S-PF-06873600 5MG
SDP
Catalog No. 502581419
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. (S)-PF-06873600 is the S enantiomer of PF-06873600 which is an inhibitor of CDK. purity: 98%
Catalog No. 50-258-1419 Supplier TARGETMOL CHEMICALS INC Supplier No. T128095MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. (S)-PF-06873600 is the S enantiomer of PF-06873600 which is an inhibitor of CDK. purity: 98%