Learn More
Cayman Chemical (S)-PFI-2 (hydrochloride), Purity: ≥95%, CAS: 1627607-88-8, 10 mg
SDP
Catalog No. 502966619
Encompass_Preferred
The inactive enantiomer of the SET7/9 inhibitor (R)-PFI-2 that may serve as a negative control
Catalog No. 50-296-6619 Supplier Cayman Chemical Supplier No. 1811910 MG
The inactive enantiomer of the SET7/9 inhibitor (R)-PFI-2 that may serve as a negative control