Learn More
Cayman Chemical S-PhenylcysteIn 10mg
SDP
Catalog No. 502988784
Encompass_Preferred
An adduct; formed by the binding of benzene oxide to cysteine residues in hemoglobin; has been found in red blood cells isolated from benzene-exposed rats and mice
Catalog No. 50-298-8784 Supplier Cayman Chemical Supplier No. 2973410 MG
An adduct; formed by the binding of benzene oxide to cysteine residues in hemoglobin; has been found in red blood cells isolated from benzene-exposed rats and mice