Learn More
Apexbio Technology LLC (S)-(-)-Phenylethanol 1445-91-6 1g
SDP
Catalog No. 5000090118
Encompass_Preferred
(S)-(-)-Phenylethanol (CAS 1445-91-6) is a chiral aromatic alcohol characterized by a stereospecific phenylethyl group Frequently employed as a synthetic intermediate or chiral building block it facilitates the investigation of asymmetric synthesis in pharmaceutical research Its defined stereochemistry makes it valuable for studying enzyme-catalyzed transformations and the molecular basis of chiral recognition including interactions with olfactory receptors Additionally (S)-(-)-Phenylethanol is applied in elucidating metabolic pathways and structure activity relationships of chiral bioactive compounds thereby supporting targeted drug design and development
Catalog No. 50-000-90118 Supplier Apexbio Technology LLC Supplier No. C66061000
(S)-(-)-Phenylethanol (CAS 1445-91-6) is a chiral aromatic alcohol characterized by a stereospecific phenylethyl group Frequently employed as a synthetic intermediate or chiral building block it facilitates the investigation of asymmetric synthesis in pharmaceutical research Its defined stereochemistry makes it valuable for studying enzyme-catalyzed transformations and the molecular basis of chiral recognition including interactions with olfactory receptors Additionally (S)-(-)-Phenylethanol is applied in elucidating metabolic pathways and structure activity relationships of chiral bioactive compounds thereby supporting targeted drug design and development