Learn More
Cayman Chemical S-PhenylmercapturIc AcId 50mg
SDP
Catalog No. 502980718
Encompass_Preferred
A metabolite of benzene; used as a biomarker for benzene exposure; urinary concentrations up to 543 UG/g creatinine have been detected
Catalog No. 50-298-0718 Supplier Cayman Chemical Supplier No. 2086950 MG
A metabolite of benzene; used as a biomarker for benzene exposure; urinary concentrations up to 543 UG/g creatinine have been detected