Learn More
AdipoGen S-Phenylmercapturic acid
SDP
Catalog No. 502560256
Encompass_Preferred
Chemical. CAS 4775-80-8. Formula C11H13NO3S. MW 239.29. Synthetic. S-Phenylmercapturic acid S-PMA is an important metabolite of benzene used as a biomarker for benzene exposure in humans. In industrial workers, urinary concentrations of up to 543 µg/g creatinine have been detected. In a study of benzene exposure through tobacco smoke inhalation in humans, smokers had a urinary concentration of 9.1 µg S-PMA/g creatinine compared with 4.8 µg S-PMA/g creatinine in non-smokers. S-PMA can be sensitively and reliably determined using capillary gas chromatographic/mass spectrometric method. Due to its sensitivity the method is suitable not only for determining concentrations relevant to occupational medicine, but also those which occur in the ecological range. Compound can be used as analytical reference material.
Catalog No. 50-256-0256 Supplier AdipoGen Supplier No. CDXP0091M100
Chemical. CAS 4775-80-8. Formula C11H13NO3S. MW 239.29. Synthetic. S-Phenylmercapturic acid S-PMA is an important metabolite of benzene used as a biomarker for benzene exposure in humans. In industrial workers, urinary concentrations of up to 543 µg/g creatinine have been detected. In a study of benzene exposure through tobacco smoke inhalation in humans, smokers had a urinary concentration of 9.1 µg S-PMA/g creatinine compared with 4.8 µg S-PMA/g creatinine in non-smokers. S-PMA can be sensitively and reliably determined using capillary gas chromatographic/mass spectrometric method. Due to its sensitivity the method is suitable not only for determining concentrations relevant to occupational medicine, but also those which occur in the ecological range. Compound can be used as analytical reference material.