Learn More
Medchemexpress LLC HY-131345A 5mg Medchemexpress, (S)-PI3Kα-IN-4 CAS:2322293-84-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY131345A5MG
Encompass_Preferred
Medchemexpress, HY-131345A 5mg (S)-PI3Kα-IN-4 CAS:2322293-84-3 (S)-PI3Kα-IN-4 is a potent inhibitor of PI3Kα, with an IC50 of 2.3 nM. (S)-PI3Kα-IN-4 shows 38.3-, 4.25-, and 4.93-fold selectivity for PI3Kα over PI3Kβ, PI3Kδ, and PI3Kγ, respectively. (S)-PI3Kα-IN-4 can be used for the research of cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1280