Learn More
Meso Scale Discovery S-PLEX Human CTLA-4 Kit
SDP
Catalog No. 5000079030
Encompass
The S-PLEX Human CTLA-4 Kit by Meso Scale Discovery is a multiplex immunoassay for measuring human CTLA-4 in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection in a 96-well plate format The kit includes capture and detection antibodies calibrators and controls for a standard curve It offers high sensitivity a broad dynamic range and low sample volume requirements testing up to 40 samples in duplicate
Catalog No. 50-000-79030 Supplier Meso Scale Discovery Supplier No. K151AGLS1
The S-PLEX Human CTLA-4 Kit by Meso Scale Discovery is a multiplex immunoassay for measuring human CTLA-4 in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection in a 96-well plate format The kit includes capture and detection antibodies calibrators and controls for a standard curve It offers high sensitivity a broad dynamic range and low sample volume requirements testing up to 40 samples in duplicate