Learn More
Meso Scale Discovery S-PLEX Human CTLA-4 Kit
SDP
Catalog No. 5000079532
Encompass
The S-PLEX Human CTLA-4 Kit by Meso Scale Discovery is a multiplex immunoassay that measures CTLA-4 levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This multiplex kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer and calibrator The assay can quantify CTLA-4 levels between 0 03 - 10 000 pg/mL and each kit provides reagents for 1 x 96-well plate
Catalog No. 50-000-79532 Supplier Meso Scale Discovery Supplier No. K151AGLS2
Add to Cart
Add to Cart
Request a Quote
The S-PLEX Human CTLA-4 Kit by Meso Scale Discovery is a multiplex immunoassay that measures CTLA-4 levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This multiplex kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer and calibrator The assay can quantify CTLA-4 levels between 0 03 - 10 000 pg/mL and each kit provides reagents for 1 x 96-well plate