Learn More
Meso Scale Discovery S-PLEX Human CTLA-4 KitQP
SDP
Catalog No. 5000079423
Encompass
The S-PLEX Human CTLA-4 Kit by Meso Scale Discovery is a multiplex immunoassay that enables the simultaneous quantitative measurement of CTLA-4 in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and is provided in a convenient 96-well plate format The kit includes a CTLA-4 calibrator and a pre-coated plate with capture antibodies enabling high sensitivity and a wide dynamic range This assay allows for the rapid and accurate quantification of CTLA-4 which plays a key role in immune regulation The kit has high lot-to-lot consistency and requires only 50 L or less sample volume per well
Catalog No. 50-000-79423 Supplier Meso Scale Discovery Supplier No. K151AGLS22
Add to Cart
Add to Cart
Request a Quote
The S-PLEX Human CTLA-4 Kit by Meso Scale Discovery is a multiplex immunoassay that enables the simultaneous quantitative measurement of CTLA-4 in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and is provided in a convenient 96-well plate format The kit includes a CTLA-4 calibrator and a pre-coated plate with capture antibodies enabling high sensitivity and a wide dynamic range This assay allows for the rapid and accurate quantification of CTLA-4 which plays a key role in immune regulation The kit has high lot-to-lot consistency and requires only 50 L or less sample volume per well