Learn More
Meso Scale Discovery S-PLEX Human Eotaxin-3 Kit
SDP
Catalog No. 5000082310
Encompass
The Meso Scale Discovery S-PLEX Human Eotaxin-3 Kit is a multiplex immunoassay that quantitatively measures human Eotaxin-3 in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium calibrators and controls The assay has a typical sensitivity of 0 08 pg/mL and an upper limit of quantification of 10 000 pg/mL This product can help researchers investigate the role of Eotaxin-3 in inflammatory diseases
Catalog No. 50-000-82310 Supplier Meso Scale Discovery Supplier No. K151Q3S2
The Meso Scale Discovery S-PLEX Human Eotaxin-3 Kit is a multiplex immunoassay that quantitatively measures human Eotaxin-3 in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium calibrators and controls The assay has a typical sensitivity of 0 08 pg/mL and an upper limit of quantification of 10 000 pg/mL This product can help researchers investigate the role of Eotaxin-3 in inflammatory diseases