Learn More
Meso Scale Discovery S-PLEX Human Eotaxin-3 KitQP
SDP
Catalog No. 5000080585
Encompass
The Meso Scale Discovery S-PLEX Human Eotaxin-3 Kit is a multiplex immunoassay that quantitatively measures human Eotaxin-3 in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and controls The assay takes approximately 3 5 hours to complete and has a lower limit of detection of 19 5 pg/mL This product enables the accurate quantification of Eotaxin-3 for research applications
Catalog No. 50-000-80585 Supplier Meso Scale Discovery Supplier No. K151Q3S22
The Meso Scale Discovery S-PLEX Human Eotaxin-3 Kit is a multiplex immunoassay that quantitatively measures human Eotaxin-3 in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and controls The assay takes approximately 3 5 hours to complete and has a lower limit of detection of 19 5 pg/mL This product enables the accurate quantification of Eotaxin-3 for research applications