Learn More
Meso Scale Discovery S-PLEX Human G-CSF Kit 25 PL
SDP
Catalog No. 5000082274
Encompass
The Meso Scale Discovery S-PLEX Human G-CSF Kit is a multiplex immunoassay kit for the quantitative measurement of granulocyte colony-stimulating factor (G-CSF) in human serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators diluent wash buffer read buffer and plate seals The kit can perform up to 94 tests in duplicate and has a testing time of 3 5 hours
Catalog No. 50-000-82274 Supplier Meso Scale Discovery Supplier No. K151K3S4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery S-PLEX Human G-CSF Kit is a multiplex immunoassay kit for the quantitative measurement of granulocyte colony-stimulating factor (G-CSF) in human serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators diluent wash buffer read buffer and plate seals The kit can perform up to 94 tests in duplicate and has a testing time of 3 5 hours