Learn More
Meso Scale Discovery S-PLEX Human G-CSF KitQP
SDP
Catalog No. 5000082338
Encompass
The Meso Scale Discovery S-PLEX Human G-CSF Kit is a multiplex immunoassay kit for the quantitative measurement of granulocyte colony-stimulating factor (G-CSF) in human serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can quantify G-CSF concentrations from 0 04-10 000 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator and controls
Catalog No. 50-000-82338 Supplier Meso Scale Discovery Supplier No. K151K3S22
The Meso Scale Discovery S-PLEX Human G-CSF Kit is a multiplex immunoassay kit for the quantitative measurement of granulocyte colony-stimulating factor (G-CSF) in human serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can quantify G-CSF concentrations from 0 04-10 000 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator and controls