Learn More
Meso Scale Discovery S-PLEX Human G-CSF KitQP
SDP
Catalog No. 5000082341
Encompass
The Meso Scale Discovery S-PLEX Human G-CSF Kit is a multiplex immunoassay that measures Granulocyte Colony Stimulating Factor (G-CSF) in human serum plasma and cell culture supernatant samples The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can quantify G-CSF over a range of 0 04-2500 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator and controls This product enables reliable quantification of G-CSF for research applications
Catalog No. 50-000-82341 Supplier Meso Scale Discovery Supplier No. K151K3S21
The Meso Scale Discovery S-PLEX Human G-CSF Kit is a multiplex immunoassay that measures Granulocyte Colony Stimulating Factor (G-CSF) in human serum plasma and cell culture supernatant samples The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can quantify G-CSF over a range of 0 04-2500 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator and controls This product enables reliable quantification of G-CSF for research applications