Learn More
Meso Scale Discovery S-PLEX Human GM-CSF Kit
SDP
Catalog No. 5000079656
Encompass
The S-PLEX Human GM-CSF Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of human granulocyte-macrophage colony-stimulating factor (GM-CSF) in serum plasma and cell culture supernatants The kit utilizes MSD s electrochemiluminescence (ECL) detection technology and provides high sensitivity a wide dynamic range and requires low sample volume This assay kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator and controls The lower limit of detection is 0 04 pg/mL and the upper limit is 1500 pg/mL
Catalog No. 50-000-79656 Supplier Meso Scale Discovery Supplier No. K151F3S1
The S-PLEX Human GM-CSF Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of human granulocyte-macrophage colony-stimulating factor (GM-CSF) in serum plasma and cell culture supernatants The kit utilizes MSD s electrochemiluminescence (ECL) detection technology and provides high sensitivity a wide dynamic range and requires low sample volume This assay kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator and controls The lower limit of detection is 0 04 pg/mL and the upper limit is 1500 pg/mL