Learn More
Meso Scale Discovery S-PLEX Human GM-CSF KitQP
SDP
Catalog No. 5000078747
Encompass
The S-Plex Human GM-CSF Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of granulocyte-macrophage colony-stimulating factor (GM-CSF) in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent calibrators and controls The electrochemiluminescent assay has a sensitivity of 0 04 pg/mL and an upper limit of quantification of 1500 pg/mL The kit can perform up to 80 tests in the provided 96-well plate
Catalog No. 50-000-78747 Supplier Meso Scale Discovery Supplier No. K151F3S22
The S-Plex Human GM-CSF Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of granulocyte-macrophage colony-stimulating factor (GM-CSF) in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent calibrators and controls The electrochemiluminescent assay has a sensitivity of 0 04 pg/mL and an upper limit of quantification of 1500 pg/mL The kit can perform up to 80 tests in the provided 96-well plate