Learn More
Meso Scale Discovery S-PLEX Human GM-CSF KitQP
SDP
Catalog No. 5000081635
Encompass
The Meso Scale Discovery S-PLEX Human GM-CSF Kit is a multiplex immunoassay that measures GM-CSF in human serum plasma and cell culture supernatant samples The kit uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume This assay measures GM-CSF levels between 0 06 - 1500 pg/mL and the total assay time is approximately 3 5 hours The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer calibrator and controls This assay allows for the quantitative measurement of GM-CSF which can aid in understanding immune system function
Catalog No. 50-000-81635 Supplier Meso Scale Discovery Supplier No. K151F3S21
The Meso Scale Discovery S-PLEX Human GM-CSF Kit is a multiplex immunoassay that measures GM-CSF in human serum plasma and cell culture supernatant samples The kit uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume This assay measures GM-CSF levels between 0 06 - 1500 pg/mL and the total assay time is approximately 3 5 hours The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer calibrator and controls This assay allows for the quantitative measurement of GM-CSF which can aid in understanding immune system function