Learn More
Meso Scale Discovery S-PLEX Human IFN- 2a KitQP
SDP
Catalog No. 5000079605
Encompass
The S-PLEX Human IFN alpha2a Kit from Meso Scale Discovery is an immunoassay kit for the quantitative measurement of human interferon alpha2a (IFN 2a) in serum plasma cell culture supernatants cell lysates and tissue homogenates The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer sample diluent calibrator and control The assay can detect IFN 2a in the range of 3 2-10 000 pg/mL and requires 2 5 hours to complete The kit includes sufficient reagents for 1 x 96-well plate
Catalog No. 50-000-79605 Supplier Meso Scale Discovery Supplier No. K151P3S22
The S-PLEX Human IFN alpha2a Kit from Meso Scale Discovery is an immunoassay kit for the quantitative measurement of human interferon alpha2a (IFN 2a) in serum plasma cell culture supernatants cell lysates and tissue homogenates The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer sample diluent calibrator and control The assay can detect IFN 2a in the range of 3 2-10 000 pg/mL and requires 2 5 hours to complete The kit includes sufficient reagents for 1 x 96-well plate