Learn More
Meso Scale Discovery S-PLEX Human IFN-α2a Kit, 5 plates, detection limit 4.9 fg/mL, dynamic range 4.9 - 67,000 fg/mL, sample types: serum, plasma, cell culture supernatant, plate type: 96-well
SDP
Catalog No. NC2046926
Encompass
S-PLEX Human IFN-α2a Kit, 5 plates, detection limit 4.9 fg/mL, dynamic range 4.9 - 67,000 fg/mL, sample types: serum, plasma, cell culture supernatant, plate type: 96-well
Catalog No. NC2046926 Supplier Meso Scale Discovery Supplier No. K151P3S2
S-PLEX Human IFN-α2a Kit, 5 plates, detection limit 4.9 fg/mL, dynamic range 4.9 - 67,000 fg/mL, sample types: serum, plasma, cell culture supernatant, plate type: 96-well