Learn More
Meso Scale Discovery S-PLEX Human IFN-B Kit 25 PL
SDP
Catalog No. 5000078940
Encompass
The S-PLEX Human IFN beta Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of human interferon beta (IFN- ) in serum plasma cell culture supernatants and other biological samples The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer sample diluent calibrators and controls The assay has a typical sensitivity of 0 08 pg/mL and an upper limit of quantification of 2500 pg/mL for IFN-
Catalog No. 50-000-78940 Supplier Meso Scale Discovery Supplier No. K151ADRS4
Add to Cart
Add to Cart
Request a Quote
The S-PLEX Human IFN beta Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of human interferon beta (IFN- ) in serum plasma cell culture supernatants and other biological samples The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer sample diluent calibrators and controls The assay has a typical sensitivity of 0 08 pg/mL and an upper limit of quantification of 2500 pg/mL for IFN-