Learn More
Meso Scale Discovery S-PLEX Human IFN-B KitQP
SDP
Catalog No. 5000079529
Encompass
The S-PLEX Human IFN beta Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of human interferon beta (IFN- ) in serum plasma cell culture supernatants and other biological samples The assay uses MSD s proprietary SECTOR Imager and SULFO-TAG labels for high sensitivity and broad dynamic range The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to perform the assay The lower limit of detection is 0 04 pg/mL and the upper limit of quantitation is 1500 pg/mL The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-79529 Supplier Meso Scale Discovery Supplier No. K151ADRS22
The S-PLEX Human IFN beta Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of human interferon beta (IFN- ) in serum plasma cell culture supernatants and other biological samples The assay uses MSD s proprietary SECTOR Imager and SULFO-TAG labels for high sensitivity and broad dynamic range The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to perform the assay The lower limit of detection is 0 04 pg/mL and the upper limit of quantitation is 1500 pg/mL The assay takes approximately 3 5 hours to complete