Learn More
Meso Scale Discovery S-PLEX Human IFN- KitQP
SDP
Catalog No. 5000081742
Encompass
The S-PLEX Human IFN gamma Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of human interferon gamma (IFN-) in serum plasma cell culture supernatants and other biological samples The kit contains a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent calibrators and controls The assay uses electrochemiluminescence (ECL) technology for high sensitivity and broad dynamic range Results can be read on MSD instruments like QuickPlex SECTOR Imager MESO QuickPlex SQ 120 and Discovery Workbench The kit has a standard curve range of 0 53-2500 pg/mL and a minimum detectable dose of 0 53 pg/mL
Catalog No. 50-000-81742 Supplier Meso Scale Discovery Supplier No. K151X9S21
The S-PLEX Human IFN gamma Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of human interferon gamma (IFN-) in serum plasma cell culture supernatants and other biological samples The kit contains a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent calibrators and controls The assay uses electrochemiluminescence (ECL) technology for high sensitivity and broad dynamic range Results can be read on MSD instruments like QuickPlex SECTOR Imager MESO QuickPlex SQ 120 and Discovery Workbench The kit has a standard curve range of 0 53-2500 pg/mL and a minimum detectable dose of 0 53 pg/mL