Learn More
Meso Scale Discovery S-PLEX Human IFN- KitQP
SDP
Catalog No. 5000082403
Encompass
The S-PLEX Human IFN gamma Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of human interferon gamma (IFN-) in serum plasma tissue culture media and other biological samples The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label assay diluent wash buffer read buffer and IFN- standard The assay has a typical sensitivity of 0 05 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-82403 Supplier Meso Scale Discovery Supplier No. K151X9S22
The S-PLEX Human IFN gamma Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of human interferon gamma (IFN-) in serum plasma tissue culture media and other biological samples The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label assay diluent wash buffer read buffer and IFN- standard The assay has a typical sensitivity of 0 05 pg/mL and an upper limit of quantification of 10 000 pg/mL