Learn More
Meso Scale Discovery S-PLEX Human IL-10 Kit
SDP
Catalog No. 5000079985
Encompass
The S-PLEX Human IL-10 Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of human interleukin 10 (IL-10) in serum plasma and cell culture supernatants The kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Diluent 2 Calibrator Blend and sample diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL
This item is not returnable. View return policy
Catalog No. 50-000-79985 Supplier Meso Scale Discovery Supplier No. K151Y2S1
The S-PLEX Human IL-10 Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of human interleukin 10 (IL-10) in serum plasma and cell culture supernatants The kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Diluent 2 Calibrator Blend and sample diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL