Learn More
Meso Scale Discovery S-PLEX Human IL-10 KitQP
SDP
Catalog No. 5000079666
Encompass
The Meso Scale Discovery S-PLEX Human IL-10 Kit is a multiplex immunoassay that measures IL-10 levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium read buffer wash buffer diluent and calibrators to generate a standard curve The assay takes 3 5 hours to complete and requires a MESO QuickPlex SQ 120 or QuickPlex SQ 120
Catalog No. 50-000-79666 Supplier Meso Scale Discovery Supplier No. K151Y2S22
The Meso Scale Discovery S-PLEX Human IL-10 Kit is a multiplex immunoassay that measures IL-10 levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium read buffer wash buffer diluent and calibrators to generate a standard curve The assay takes 3 5 hours to complete and requires a MESO QuickPlex SQ 120 or QuickPlex SQ 120