Learn More
Meso Scale Discovery S-PLEX Human IL-12p70 KitQP
SDP
Catalog No. 5000082007
Encompass
The Meso Scale Discovery S-PLEX Human IL-12p70 Kit is an immunoassay using electrochemiluminescence (ECL) for measuring human interleukin 12 (IL-12) in serum plasma and cell culture supernatants It employs a sandwich format with pre-coated antibodies on multi-array plates Samples and calibrators are added and IL-12 binds to the immobilized antibody After washing a ruthenylated detection antibody binds to IL-12 A read buffer enables ECL detection with MSD instruments The kit includes a plate detection antibody read buffer wash buffer blocker calibrators and positive control
Catalog No. 50-000-82007 Supplier Meso Scale Discovery Supplier No. K151G3S22
The Meso Scale Discovery S-PLEX Human IL-12p70 Kit is an immunoassay using electrochemiluminescence (ECL) for measuring human interleukin 12 (IL-12) in serum plasma and cell culture supernatants It employs a sandwich format with pre-coated antibodies on multi-array plates Samples and calibrators are added and IL-12 binds to the immobilized antibody After washing a ruthenylated detection antibody binds to IL-12 A read buffer enables ECL detection with MSD instruments The kit includes a plate detection antibody read buffer wash buffer blocker calibrators and positive control