Learn More
Meso Scale Discovery S-PLEX Human IL-13 KitQP
SDP
Catalog No. 5000079727
Encompass
The Meso Scale Discovery S-PLEX Human IL-13 Kit is a multiplex immunoassay for measuring IL-13 in human serum plasma and cell culture supernatants It includes a 96-well plate with IL-13 capture antibodies detection antibodies with an electrochemiluminescent label calibrators diluent read and wash buffers and plate seals The detection range is 0 03-2500 pg/mL using 50 L of sample and takes about 3 5 hours Compatible with MSD QuickPlex SQ 120 MESO QuickPlex SQ 120 and SECTOR S 600 it offers high sensitivity small sample volume and minimal cross-reactivity
Catalog No. 50-000-79727 Supplier Meso Scale Discovery Supplier No. K151Y9S22
The Meso Scale Discovery S-PLEX Human IL-13 Kit is a multiplex immunoassay for measuring IL-13 in human serum plasma and cell culture supernatants It includes a 96-well plate with IL-13 capture antibodies detection antibodies with an electrochemiluminescent label calibrators diluent read and wash buffers and plate seals The detection range is 0 03-2500 pg/mL using 50 L of sample and takes about 3 5 hours Compatible with MSD QuickPlex SQ 120 MESO QuickPlex SQ 120 and SECTOR S 600 it offers high sensitivity small sample volume and minimal cross-reactivity