Learn More
Meso Scale Discovery S-PLEX Human IL-17A KitQP
SDP
Catalog No. 5000081976
Encompass
The Meso Scale Discovery S-PLEX Human IL-17A Kit is a multiplex immunoassay for measuring IL-17A in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG Read Buffer Diluent 2 and calibrators and controls for a standard curve The assay takes about 3 5 hours and requires a MESO QuickPlex SQ 120 or MSD MULTI-ARRAY instrument
Catalog No. 50-000-81976 Supplier Meso Scale Discovery Supplier No. K151C3S22
Add to Cart
Add to Cart
The Meso Scale Discovery S-PLEX Human IL-17A Kit is a multiplex immunoassay for measuring IL-17A in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG Read Buffer Diluent 2 and calibrators and controls for a standard curve The assay takes about 3 5 hours and requires a MESO QuickPlex SQ 120 or MSD MULTI-ARRAY instrument