Learn More
Meso Scale Discovery S-PLEX Human IL-1B Kit 25 PL
SDP
Catalog No. 5000080357
Encompass
The S-PLEX Human IL-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of human interleukin 1 beta (IL-1 ) in serum plasma and cell culture supernatants The kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume The assay has high specificity for IL-1 with no cross-reactivity to other cytokines The kit includes a 96-well plate coated with capture antibody detection antibody read buffer diluent calibrator and control The kit can perform up to 94 tests in duplicate
Catalog No. 50-000-80357 Supplier Meso Scale Discovery Supplier No. K151ADSS4
Add to Cart
Add to Cart
Request a Quote
The S-PLEX Human IL-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of human interleukin 1 beta (IL-1 ) in serum plasma and cell culture supernatants The kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume The assay has high specificity for IL-1 with no cross-reactivity to other cytokines The kit includes a 96-well plate coated with capture antibody detection antibody read buffer diluent calibrator and control The kit can perform up to 94 tests in duplicate