Learn More
Meso Scale Discovery S-PLEX Human IL-1B KitQP
SDP
Catalog No. 5000078944
Encompass
The Meso Scale Discovery S-PLEX Human IL-1 Kit is a multiplex immunoassay that measures IL-1 in human serum plasma and cell culture supernatant samples The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators diluent wash buffer read buffer and plate seals The assay takes approximately 3 5 hours to complete and has a lower limit of detection of 0 04 pg/mL
Catalog No. 50-000-78944 Supplier Meso Scale Discovery Supplier No. K151ADSS21
The Meso Scale Discovery S-PLEX Human IL-1 Kit is a multiplex immunoassay that measures IL-1 in human serum plasma and cell culture supernatant samples The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators diluent wash buffer read buffer and plate seals The assay takes approximately 3 5 hours to complete and has a lower limit of detection of 0 04 pg/mL