Learn More
Meso Scale Discovery S-PLEX Human IL-1B KitQP
SDP
Catalog No. 5000080356
Encompass
The S-PLEX Human IL-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of human interleukin 1 beta (IL-1 ) in serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators diluent read buffer wash buffer and plate seals The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 1500 pg/mL
Catalog No. 50-000-80356 Supplier Meso Scale Discovery Supplier No. K151ADSS22
The S-PLEX Human IL-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of human interleukin 1 beta (IL-1 ) in serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators diluent read buffer wash buffer and plate seals The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 1500 pg/mL