Learn More
Meso Scale Discovery S-PLEX Human IL-2 Kit
SDP
Catalog No. 5000079826
Encompass
The Meso Scale Discovery S-PLEX Human IL-2 Kit is a multiplex immunoassay that measures human interleukin-2 (IL-2) in serum plasma and cell culture supernatant samples The assay uses electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent The assay takes approximately 3 5 hours to complete after a 2 hour incubation
Catalog No. 50-000-79826 Supplier Meso Scale Discovery Supplier No. K151Z2S1
The Meso Scale Discovery S-PLEX Human IL-2 Kit is a multiplex immunoassay that measures human interleukin-2 (IL-2) in serum plasma and cell culture supernatant samples The assay uses electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent The assay takes approximately 3 5 hours to complete after a 2 hour incubation