Learn More
Meso Scale Discovery S-PLEX Human IL-2 KitQP
SDP
Catalog No. 5000081848
Encompass
The Meso Scale Discovery S-PLEX Human IL-2 Kit is a multiplex immunoassay that measures human interleukin 2 (IL-2) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium read buffer wash buffer calibrator and controls The assay has a typical sensitivity of 0 06 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-81848 Supplier Meso Scale Discovery Supplier No. K151Z2S22
The Meso Scale Discovery S-PLEX Human IL-2 Kit is a multiplex immunoassay that measures human interleukin 2 (IL-2) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium read buffer wash buffer calibrator and controls The assay has a typical sensitivity of 0 06 pg/mL and an upper limit of quantification of 10 000 pg/mL