Learn More
Meso Scale Discovery S-PLEX Human IL-21 Kit
SDP
Catalog No. 5000079494
Encompass
The Meso Scale Discovery S-PLEX Human IL-21 Kit is a multiplex immunoassay for measuring human interleukin 21 (IL-21) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies ruthenium-labeled detection antibodies buffers calibrator and controls The assay takes about 3 5 hours and requires a MESO QuickPlex SQ 120 or SQ 120Y instrument for analysis enabling accurate IL-21 quantitation
Catalog No. 50-000-79494 Supplier Meso Scale Discovery Supplier No. K151ADNS1
The Meso Scale Discovery S-PLEX Human IL-21 Kit is a multiplex immunoassay for measuring human interleukin 21 (IL-21) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies ruthenium-labeled detection antibodies buffers calibrator and controls The assay takes about 3 5 hours and requires a MESO QuickPlex SQ 120 or SQ 120Y instrument for analysis enabling accurate IL-21 quantitation