Learn More
Meso Scale Discovery S-PLEX Human IL-21 Kit 25 PL
SDP
Catalog No. 5000079425
Encompass
The Meso Scale Discovery S-PLEX Human IL-21 Kit is a multiplex immunoassay for measuring IL-21 in human serum plasma and cell culture supernatant It uses electrochemiluminescent detection on Meso Scale Discovery instruments The kit includes a 96-well plate with IL-21 capture antibodies detection antibodies with electrochemiluminescent tags calibrator diluent wash buffer read buffer and a lyophilized calibrator with known IL-21 concentration The assay s sensitivity is 0 3 pg/mL with an upper quantification limit of 2500 pg/mL
Catalog No. 50-000-79425 Supplier Meso Scale Discovery Supplier No. K151ADNS4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery S-PLEX Human IL-21 Kit is a multiplex immunoassay for measuring IL-21 in human serum plasma and cell culture supernatant It uses electrochemiluminescent detection on Meso Scale Discovery instruments The kit includes a 96-well plate with IL-21 capture antibodies detection antibodies with electrochemiluminescent tags calibrator diluent wash buffer read buffer and a lyophilized calibrator with known IL-21 concentration The assay s sensitivity is 0 3 pg/mL with an upper quantification limit of 2500 pg/mL