Learn More
Meso Scale Discovery S-PLEX Human IL-21 KitQP
SDP
Catalog No. 5000079535
Encompass
The Meso Scale Discovery S-PLEX Human IL-21 Kit is a multiplex immunoassay that measures human interleukin 21 (IL-21) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium read buffer wash buffer blocker solutions calibrators and controls The assay takes approximately 3 5 hours to complete and has a lower limit of detection of 19 5 pg/mL for IL-21
Catalog No. 50-000-79535 Supplier Meso Scale Discovery Supplier No. K151ADNS22
The Meso Scale Discovery S-PLEX Human IL-21 Kit is a multiplex immunoassay that measures human interleukin 21 (IL-21) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium read buffer wash buffer blocker solutions calibrators and controls The assay takes approximately 3 5 hours to complete and has a lower limit of detection of 19 5 pg/mL for IL-21