Learn More
Meso Scale Discovery S-PLEX Human IL-22 Kit, 5 each, 2.2 fg/mL sensitivity, dynamic range 2.2 - 35,000 fg/mL, compatible with 96-well SECTOR or QuickPlex plates
SDP
Catalog No. NC2861735
Encompass
S-PLEX Human IL-22 Kit, 5 each, 2.2 fg/mL sensitivity, dynamic range 2.2 - 35,000 fg/mL, compatible with 96-well SECTOR or QuickPlex plates
Catalog No. NC2861735 Supplier Meso Scale Discovery Supplier No. K151H3S2
S-PLEX Human IL-22 Kit, 5 each, 2.2 fg/mL sensitivity, dynamic range 2.2 - 35,000 fg/mL, compatible with 96-well SECTOR or QuickPlex plates