Learn More
Meso Scale Discovery S-PLEX Human IL-22 Kit 25 PL
SDP
Catalog No. 5000079155
Encompass
The Meso Scale Discovery S-PLEX Human IL-22 Kit is a multiplex immunoassay that measures IL-22 in human serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer Diluent 2 Calibrator Blend and Wash Buffer The assay takes 3 5 hours to complete and has a lower limit of detection of 0 23 pg/mL The kit can perform up to 80 tests and has a 12-month shelf life when stored at 2-8 C
Catalog No. 50-000-79155 Supplier Meso Scale Discovery Supplier No. K151H3S4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery S-PLEX Human IL-22 Kit is a multiplex immunoassay that measures IL-22 in human serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer Diluent 2 Calibrator Blend and Wash Buffer The assay takes 3 5 hours to complete and has a lower limit of detection of 0 23 pg/mL The kit can perform up to 80 tests and has a 12-month shelf life when stored at 2-8 C