Learn More
Meso Scale Discovery S-PLEX Human IL-22 KitQP
SDP
Catalog No. 5000080793
Encompass
The Meso Scale Discovery S-PLEX Human IL-22 Kit is a multiplex immunoassay that measures IL-22 in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium and calibrators and controls needed to quantify IL-22 levels This assay can help researchers investigate the role of IL-22 in autoimmunity inflammation and tissue regeneration
Catalog No. 50-000-80793 Supplier Meso Scale Discovery Supplier No. K151H3S21
The Meso Scale Discovery S-PLEX Human IL-22 Kit is a multiplex immunoassay that measures IL-22 in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium and calibrators and controls needed to quantify IL-22 levels This assay can help researchers investigate the role of IL-22 in autoimmunity inflammation and tissue regeneration