Learn More
Meso Scale Discovery S-PLEX Human IL-3 Kit 25 PL
SDP
Catalog No. 5000079457
Encompass
The S-PLEX Human IL-3 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human interleukin 3 (IL-3) in serum plasma and cell culture supernatants using electrochemiluminescence (ECL) detection technology The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 08 pg/mL and an upper limit of quantification of 2500 pg/mL for human IL-3 The total assay time is 3 5 hours
Catalog No. 50-000-79457 Supplier Meso Scale Discovery Supplier No. K151ADQS4
Add to Cart
Add to Cart
Request a Quote
The S-PLEX Human IL-3 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human interleukin 3 (IL-3) in serum plasma and cell culture supernatants using electrochemiluminescence (ECL) detection technology The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 08 pg/mL and an upper limit of quantification of 2500 pg/mL for human IL-3 The total assay time is 3 5 hours