Learn More
Meso Scale Discovery S-PLEX Human IL-3 Kit
SDP
Catalog No. 5000078947
Encompass
The Meso Scale Discovery Human IL-3 Singleplex Assay Kit is a sensitive immunoassay for measuring human interleukin 3 (IL-3) in various biological samples It uses electrochemiluminescence (ECL) detection for accurate reproducible results with high sensitivity and a broad dynamic range The assay requires minimal sample volume and hands-on time suitable for biomarker discovery cell signaling research and monitoring disease progression The kit includes a 96-well plate with capture antibody detection antibody read buffer wash buffer calibrator and controls
Catalog No. 50-000-78947 Supplier Meso Scale Discovery Supplier No. K151ADQS1
Add to Cart
Add to Cart
The Meso Scale Discovery Human IL-3 Singleplex Assay Kit is a sensitive immunoassay for measuring human interleukin 3 (IL-3) in various biological samples It uses electrochemiluminescence (ECL) detection for accurate reproducible results with high sensitivity and a broad dynamic range The assay requires minimal sample volume and hands-on time suitable for biomarker discovery cell signaling research and monitoring disease progression The kit includes a 96-well plate with capture antibody detection antibody read buffer wash buffer calibrator and controls