Learn More
Meso Scale Discovery S-PLEX Human IL-3 Kit
SDP
Catalog No. 5000079583
Encompass
The Meso Scale Discovery Human IL-3 Singleplex Assay is a highly sensitive immunoassay that quantitatively measures human interleukin 3 (IL-3) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides accurate reproducible results with a lower limit of detection of 0 08 pg/mL The assay requires a small sample volume of 50 L and results can be read in as little as 3 5 hours using Meso Scale Discovery instrumentation This IL-3 assay provides a rapid ultra-sensitive method for quantifying IL-3 levels in human biological samples
Catalog No. 50-000-79583 Supplier Meso Scale Discovery Supplier No. K151ADQS2
The Meso Scale Discovery Human IL-3 Singleplex Assay is a highly sensitive immunoassay that quantitatively measures human interleukin 3 (IL-3) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides accurate reproducible results with a lower limit of detection of 0 08 pg/mL The assay requires a small sample volume of 50 L and results can be read in as little as 3 5 hours using Meso Scale Discovery instrumentation This IL-3 assay provides a rapid ultra-sensitive method for quantifying IL-3 levels in human biological samples